Tin, Oil Based Standard Solution, Specpure®, Sn 1000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
014206-18
AA14206-18EA
67.5
USD
AA14206-18
Tin, Oil Based Standard Solution, Specpure®, Sn 1000 µg/g, Thermo Scientific
Standards
Cationic Standards
Tin Standards
Physical Form: Liquid
MDL No.: MFCD00133862
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...