Vanadium, Oil based Standard Solution, Specpure®, V 1000 µg/g, Thermo Scientific

Supplier: Thermo Scientific Chemicals

014203-18
AA14203-18EA 67.5 USD
AA14203-18
Vanadium, Oil based Standard Solution, Specpure®, V 1000 µg/g, Thermo Scientific
Standards Cationic Standards Vanadium Standards

Physical Form: Liquid
MDL No.: MFCD00011453

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR