Zinc, Oil Based Standard Solution, Specpure®, Zn 1000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
014200-18
AA14200-18EA
79.6
USD
AA14200-18
Zinc, Oil Based Standard Solution, Specpure®, Zn 1000 µg/g, Thermo Scientific
Standards
Cationic Standards
Zinc Standards
Physical Form: Liquid
MDL No.: MFCD00011291
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...