Silver, Oil Based Standard Solution, Specpure®, Ag 1000 µg/g, Thermo Scientific
Supplier: Thermo Scientific Chemicals
014199-18
AA14199-18EA
82.7
USD
AA14199-18
Silver, Oil Based Standard Solution, Specpure®, Ag 1000 µg/g, Thermo Scientific
Standards
Cationic Standards
Silver Standards
Physical Form: Liquid
MDL No.: MFCD00003397
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...