Tributylphosphine 94%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tri-n-butylphosphine, TBP

014114-18 014114-30 014114-95
AA14114-18EA 48.8 USD
AA14114-18 AA14114-30 AA14114-95
Tributylphosphine 94%
Tributylphosphine

For cleavage of disulfide bonds, as a polymerization crosslinking catalyst, as an organic intermediate, and in fuels


Purity: 94% CAS No.: 998-40-3 Molecular Formula: [CH3(CH2)3]3P Formula Weight: 202.32 Flash Point: 93°(199°F) Physical Form: Liquid Density: 0.820 Refractive Index: 1.4620 Sensitivity: Air and Moisture Sensitive Odor: Unpleasant Melting Point: -65° Boiling Point: 240° Solubility: Insoluble in water. Miscible with ether, methanol, ethanol and benzene Beilstein Reference: 1738261 UN No.: 3254 MDL No.: MFCD00009462


For cleavage of disulfide bonds, as a polymerization crosslinking catalyst, as an organic intermediate, and in fuels

Formula: C₁₂H₂₇P
Boiling Pt: 240 °C
Melting Pt: –65 °C
Density: 0.820
Flash Pt: >93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009462
CAS Number: 998-40-3
EINECS: 213-651-2
UN: 3254
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR