Cyclopentadienylthallium

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Thallium(I) cyclopentadienide

013958-06 013958-14
AA13958-06EA 94 USD
AA13958-06 AA13958-14 AA13958-03
Cyclopentadienylthallium
Cyclopentadienylthallium

MDL: MFCD00134141


CAS No.: 34822-90-7 Molecular Formula: C5H5Tl Formula Weight: 269.47 Physical Form: Powder Sensitivity: Air Sensitive Melting Point: dec. UN No.: 1707 MDL No.: MFCD00134141

Formula: C₅H₅Tl
Boiling Pt: 90…110 °C (1 mmHg)
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00134141
CAS Number: 34822-90-7
EINECS: 252-229-2
UN: 1707
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR