Terbium, plasma standard solution, Specpure®, Tb 1000 µg/ml
Supplier: Thermo Scientific Chemicals
013860-AE
013860-AP
AA13860-AEEA
92.53
USD
AA13860-AE
AA13860-AP
Terbium, plasma standard solution, Specpure®, Tb 1000 µg/ml
Standards
Cationic Standards
Terbium Standards
MDL: MFCD00011256
Molecular Formula: Tb4O7 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011256
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...