Specpure® Sulfur, Plasma Standard Solution, S 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
013842-AD
013842-AE
013842-AP
AA13842-ADEA
55.73
USD
AA13842-AD
AA13842-AE
AA13842-AP
Specpure® Sulfur, Plasma Standard Solution, S 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Sulfur standards
Molecular Formula: (NH4)2SO4 in H2O
Physical Form: Liquid
MDL No.: MFCD00085316
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...