Sodium hydrogen sulfate monohydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

013799-36 013799-A1 013799-A7
AA13799-36EA 116.24 USD
AA13799-36 AA13799-A1 AA13799-A7
Sodium hydrogen sulfate monohydrate, REacton®, Reagent Grade
Sodium hydrogen sulphate monohydrate

MDL: MFCD00149210


CAS No.: 10034-88-5 Molecular Formula: NaHSO4.H2O Formula Weight: 138.07 (120.06anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Colorless Melting Point: ca 183° dec. Merck Reference: 14,8586 UN No.: 3260 MDL No.: MFCD00149210

Formula: NaHSO₄·H₂O
Melting Pt: ∼183 °C (dec.)
Density: 2.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149210
CAS Number: 10034-88-5
EINECS: 231-665-7
UN: 3260
ADR: 8,III
Merck Index: 14,8586

Order Now

Specification Test Results

Acidity as H2SO3 35.0-36.5
Insoluble matter and ammonium hydroxide precipitate 0.005
Cl and Br 0.001
PO3 0.001%
As 0.0001%
Ca and Mg 0.005%
Heavy Metals (as Pb) 0.0005%
Fe 0.00001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR