Sodium hydrogen sulfate monohydrate, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00149210
CAS No.: 10034-88-5
Molecular Formula: NaHSO4.H2O
Formula Weight: 138.07 (120.06anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: Colorless
Melting Point: ca 183° dec.
Merck Reference: 14,8586
UN No.: 3260
MDL No.: MFCD00149210
Formula:
NaHSO₄·H₂O Melting Pt: ∼183 °C (dec.) Density: 2.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149210 CAS Number: 10034-88-5 EINECS: 231-665-7 UN: 3260 ADR: 8,III Merck Index: 14,8586 |
Specification Test Results
Acidity as H2SO3 | 35.0-36.5 |
Insoluble matter and ammonium hydroxide precipitate | 0.005 |
Cl and Br | 0.001 |
PO3 | 0.001% |
As | 0.0001% |
Ca and Mg | 0.005% |
Heavy Metals (as Pb) | 0.0005% |
Fe | 0.00001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...