Chromium ≥99.95% (metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Warning

013796-22 013796-36 013796-14
AA13796-22EA 270 USD
AA13796-22 AA13796-36 AA13796-14
Chromium ≥99.95% (metals basis), powder -200 mesh
Chromium

MDL: MFCD00010944


Purity: 99.95% CAS No.: 7440-47-3 Appearance: Silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00010944

Formula: Cr
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR