Zinc chloride, ultra dry ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

013694-18 013694-09
AA13694-18EA 327 USD
AA13694-18 AA13694-09
Zinc chloride, ultra dry ≥99.99% (metals basis)
Zinc chloride

MDL: MFCD00011295 Fieser: 1,1289 13,349 15,368 16,391 18,410 19,409 20,439 21,493


Purity: 99.99% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn Formula Weight: 136.29 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.91 Melting Point: 290° Boiling Point: 732° Merck Reference: 14,10132 UN No.: 2331 MDL No.: MFCD00011295

Formula: ZnCl₂
Boiling Pt: 732 °C
Melting Pt: 290 °C
Density: 2.91
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 14,10132

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR