Sodium iodide, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
013604-06
013604-14
013604-22
AA13604-06EA
88.4
USD
AA13604-06
AA13604-14
AA13604-22
Sodium iodide, ultra dry ≥99.999% (metals basis)
Sodium iodide
Purity: 99.999%
CAS No.: 7681-82-5
Molecular Formula: NaI
Formula Weight: 149.89
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.67
Refractive Index: 1.7745
Sensitivity: Light Sensitive and Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 651°
Boiling Point: 1300-1304°
Merck Reference: 14,8631
MDL No.: MFCD00003532
Formula:
NaI MW: 149.89 g/mol Boiling Pt: 1300 Melting Pt: 662 Density: 3.67 Storage Temperature: Ambient |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 EINECS: 231-679-3 UN: 3077 ADR: 9 III Merck Index: 14,8631 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...