Sodium hydroxide, anhydrous ≥97.0% ACS low chloride
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
In chemical manufacture, rayon and cellophane, as a neutralizing agent in petroleum refining, in pulp, paper detergents and textiles, and as a lab reagent. NaOH solutions precipitate most metals as hydroxides from aqueous solutions of the metal salts
Purity: 97.0% min
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Physical Form: Pellets
Density: 2.13
pH: 14
Melting Point: 318°
Boiling Point: 1390°
Solubility: Soluble in water, alcohol and glycerol
Merck Reference: 14,8627
UN No.: 1823
MDL No.: MFCD00003548
In chemical manufacture, rayon and cellophane, as a neutralizing agent in petroleum refining, in pulp, paper detergents and textiles, and as a lab reagent. NaOH solutions precipitate most metals as hydroxides from aqueous solutions of the metal salts
Formula:
NaOH Boiling Pt: 1390 °C Melting Pt: 318 °C Density: 2.13 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,8627 |
Specification Test Results
Cl | 0.005% |
N | 0.1% |
PO9 | 0.001% |
SO3 | 0.003% |
Heavy Metals (as Ag) | 0.002% |
Fe | 0.001% |
Hg | 0.1 ppm |
Ca | 0.005% |
MG | 0.002% |
Ni | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...