Ammonium iron (II) sulfate hexahydrate 98.5-101.5% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate
Polymerization catalyst, photography
Purity: 98.5-101.5%
CAS No.: 7783-85-9
Molecular Formula: H8FeN2O8S2.6H2O
Formula Weight: 392.13 (284.04anhy)
Physical Form: Crystalline
Density: 1.864
Appearance: Pale bluish green
Odor: Odorless
Melting Point: ca 100° dec.
Solubility: Soluble in water. Insoluble in alcohol
Merck Reference: 14,521
MDL No.: MFCD00150530
Polymerization catalyst, photography
Formula:
(NH₄)₂Fe(SO₄)₂·6H₂O Melting Pt: ∼ 100 °C (dec.) Density: 1.864 Storage Temperature: Ambient |
MDL Number:
MFCD00150530 CAS Number: 7783-85-9 EINECS: 233-382-4 Merck Index: 14,521 |
Specification Test Results
Insoluble matter | 0.01% |
PO3 | 0.003% |
Ca | 0.005% |
Cu | 0.003% |
MG | 0.002% |
Ferric ion (Fe??) | 0.01% |
Mn | 0.0001 |
K | 0.00002 |
NA | 0.0002 |
Zn | 0.003% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...