Ammonium iron (II) sulfate hexahydrate 98.5-101.5% ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: di-Ammonium iron (II) sulfate hexahydrate, Ferrous Ammonium sulphate Hexahydrate, Ferrous ammonium sulfate hexahydrate, Ferrous Ammonium sulfate Hexahydrate, di-Ammonium iron (II) sulphate hexahydrate, Ferrous ammonium sulphate hexahydrate, Ammonium iron(Ⅱ) sulphate hexahydrate, Ammonium iron(Ⅱ) sulfate hexahydrate, Ammonium ferrous sulphate hexahydrate, Ammonium iron(II) sulphate hydrate, Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate hydrate

013448-A3 013448-36
AA13448-A3EA 231.15 USD
AA13448-A3 AA13448-36
Ammonium iron (II) sulfate hexahydrate 98.5-101.5% ACS
Ammonium iron (II) sulphate hexahydrate

Polymerization catalyst, photography


Purity: 98.5-101.5% CAS No.: 7783-85-9 Molecular Formula: H8FeN2O8S2.6H2O Formula Weight: 392.13 (284.04anhy) Physical Form: Crystalline Density: 1.864 Appearance: Pale bluish green Odor: Odorless Melting Point: ca 100° dec. Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,521 MDL No.: MFCD00150530


Polymerization catalyst, photography

Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
Melting Pt: ∼ 100 °C (dec.)
Density: 1.864
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-382-4
Merck Index: 14,521

Order Now

Specification Test Results

Insoluble matter 0.01%
PO3 0.003%
Ca 0.005%
Cu 0.003%
MG 0.002%
Ferric ion (Fe??) 0.01%
Mn 0.0001
K 0.00002
NA 0.0002
Zn 0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR