Sodium borohydride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate

013432-22 013432-36 013432-57
AA13432-22EA 150.25 USD
AA13432-22 AA13432-36 AA13432-58
Sodium borohydride 98%
Sodium borohydride

MDL: MFCD00003518 Fieser: 1,1049 15,290 16,304 17,314 18,326 19,307 20,344 21,397


Purity: 98% CAS No.: 16940-66-2 Molecular Formula: NaBH4 Formula Weight: 37.83 Physical Form: Powder Density: 1.074 Appearance: White Odor: Weak Melting Point: 400° dec. Merck Reference: 14,8592 UN No.: 1426 MDL No.: MFCD00003518

Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 400 (dec.)
Density: 1.074
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
EINECS: 241-004-4
UN: 1426
ADR: 4.3 I
Merck Index: 14,8592

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR