Lithium hydroxide monohydrate LiOH ≥ 56,5%

Supplier: Thermo Scientific Chemicals
Danger

013409-18 013409-36 013409-A3
AA13409-18EA 34.59 USD
AA13409-18 AA13409-36 AA13409-A3
Lithium hydroxide monohydrate LiOH ≥ 56,5%
Lithium hydroxide monohydrate

MDL: MFCD00149772 Fieser: 18,217 19,198 20,222


CAS No.: 1310-66-3 Molecular Formula: LiOH.H2O Formula Weight: 41.96 (23.95anhy) Physical Form: Powder Density: 1.51 Sensitivity: Air Sensitive and Hygroscopic Merck Reference: 14,5534 UN No.: 2680 MDL No.: MFCD00149772

Formula: LiOH·1H₂O
Boiling Pt: 920 °C (1013 hPa)
Melting Pt: 462 °C
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 14,5534

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR