Gold ≥99.99% (metals basis), slug, Premion®, Length 6.35 mm (0.25 in), Diameter 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

013394-KG 013394-KM 013394-KJ
AA13394-KGEA 278 USD
AA13394-KG AA13394-KM AA13394-KJ
Gold ≥99.99% (metals basis), slug, Premion®, Length 6.35 mm (0.25 in), Diameter 3.175 mm (0.125 in)
Gold

MDL: MFCD00003436


Purity: 99.99% CAS No.: 7440-57-5 Physical Form: ≈0.97g/slug Odor: Odorless MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR