Silicon dioxide ≥99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

013024-A3 013024-A9
AA13024-A3EA 72.6 USD
AA13024-A3 AA13024-A9
Silicon dioxide ≥99.5%
Silicium dioxide

In the manufacture of glass, ceramics and abrasives, in water filtration, in cosmetics, pharmaceuticals and rubber, as a thermal insulator.


Silica is used in the manufacture of glass for windows, drinking glasses, beverage bottles, and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware, and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.


98.5-99% microcrystalline silica in the form of quartzIt is a very stable compound of silicon.

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: 1710 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 238-878-4
Merck Index: 14,8493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR