Titanium(II) hydride ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Titanium hydride, Titanium dihydride
012857-06
012857-22
012857-36
012857-D3
AA12857-06EA
37.9
USD
AA12857-06
AA12857-22
AA12857-36
AA12857-D3
Titanium(II) hydride ≥99% (metals basis)
Titanium (II) hydride
Powder, 325 mesh. Moisture sensitive. Purity based on metal contaminates only.
Purity: 99%
CAS No.: 7704-98-5
Molecular Formula: H2Ti
Formula Weight: 49.92
Physical Form: -325 Mesh Powder
Density: 3.9
Appearance: Dark grey
Odor: Odorless
Melting Point: 450° dec.
Merck Reference: 14,9473
UN No.: 1871
MDL No.: MFCD00151301
Formula:
H₂Ti Melting Pt: 450 °C (dec.) Density: 3.9 Storage Temperature: Ambient |
MDL Number:
MFCD00151301 CAS Number: 7704-98-5 EINECS: 231-726-8 UN: 1871 ADR: 4.1,II Merck Index: 14,9473 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...