Sodium selenate, anhydrous ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

012613-18 012613-30 012613-06 012613-A1
AA12613-18EA 101.58 USD
AA12613-18 AA12613-30 AA12613-06 AA12613-A1
Sodium selenate, anhydrous ≥99.8% (metals basis)
Sodium selenate

Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.


Stable under alkaline and oxidizing conditions.


As a reagent

Formula: Na₂O₄Se
Melting Pt: 32 °C trans.
Density: 3.213
Storage Temperature: Ambient
MDL Number: MFCD00003490
CAS Number: 13410-01-0
EINECS: 236-501-8
UN: 2630
ADR: 6.1,I
Merck Index: 14,8671

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR