Sodium dichromate dihydrate ≥99%, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00149166
Soluble in water
Fieser: 1,1059 3,265
CAS No.: 7789-12-0
Molecular Formula: Cr2Na2O7.2H2O
Formula Weight: 298.00 (261.97anhy)
Physical Form: Crystalline
Density: 2.348
Sensitivity: Hygroscopic
Appearance: Red
Odor: Odorless
Melting Point: 100° -2H2O
Solubility: Soluble in water
Merck Reference: 14,8609
UN No.: 3087
MDL No.: MFCD00149166
Formula:
Na₂Cr₂O₇·2H₂O Melting Pt: 100 °C (-2H₂O) Density: 2.348 Storage Temperature: Ambient |
MDL Number:
MFCD00149166 CAS Number: 7789-12-0 EINECS: 234-190-3 UN: 3087 ADR: 5.1,II Merck Index: 14,8609 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...