Potassium chromate ≥99.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

012610-36 012610-22
AA12610-36EA 216.13 USD
AA12610-36 AA12610-22
Potassium chromate ≥99.0% ACS
Potassium chromate

In finishing leather, rust proofing of metals, textile mordant, as an analytical lab reagent


Purity: 99.0% min CAS No.: 7789-00-6 Molecular Formula: K2CrO4 Formula Weight: 194.19 Physical Form: Crystalline Density: 2.73 Appearance: Yellow Odor: Odorless Melting Point: 975° Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,7622 UN No.: 3087 MDL No.: MFCD00011368


In finishing leather, rust proofing of metals, textile mordant, as an analytical lab reagent

Formula: K₂CrO₄
MW: 194.19 g/mol
Boiling Pt: 1000
Melting Pt: 985
Density: 2.73
Storage Temperature: Ambient
MDL Number: MFCD00011368
CAS Number: 7789-00-6
EINECS: 232-140-5
UN: 3087
ADR: 5.1 III
Merck Index: 14,7622

Order Now

Specification Test Results

Insoluble matter 0.005%
Cl 0.005%
pH of a 5% solution 8.6-9.8 at 25°
SO3 0.03%
Ca 0.005%
NA 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR