Gold ≥99.99% (metals basis), rods, Premion®, Diameter 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

012555-HF
AA12555-HFEA 1640 USD
AA12555-HF
Gold ≥99.99% (metals basis), rods, Premion®, Diameter 6.35 mm (0.25 in)
Gold

MDL: MFCD00003436


Purity: 99.99% CAS No.: 7440-57-5 Physical Form: ≈6.1g/10mm Odor: Odorless MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR