Triphenylbismuth 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bismuth triphenyle, Triphenylbismuthine
Volatile organometallic used in the preparation of thin films of superconducting oxides by CVD
Purity: 99+%
CAS No.: 603-33-8
Molecular Formula: (C6H5)3Bi
Formula Weight: 440.30
Physical Form: Powder
Density: 1.585
Appearance: White to off-white
Melting Point: 76-81°
Boiling Point: 242°/14mm
Solubility: Slightly soluble in alcohol. Soluble in ether, acetone and chloroform. Insoluble in water
MDL No.: MFCD00014064
Volatile organometallic used in the preparation of thin films of superconducting oxides by CVD
Formula:
C₁₈H₁₅Bi Boiling Pt: 242 °C (14 mmHg) Melting Pt: 76…81 °C Density: 1,585 Storage Temperature: Ambient |
MDL Number:
MFCD00014064 CAS Number: 603-33-8 EINECS: 210-033-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...