Indium (III) oxide ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Indium trioxide, Diindium trioxide
012544-09
012544-18
012544-30
AA12544-09EA
84.7
USD
AA12544-09
AA12544-18
AA12544-30
Indium (III) oxide ≥99.99% (metals basis)
Indium (III) oxide
MDL: MFCD00011060
Purity: 99.99%
CAS No.: 1312-43-2
Molecular Formula: In2O3
Formula Weight: 277.64
Physical Form: -325 Mesh Powder
Density: 7.18
Melting Point: 850° volat.
Merck Reference: 14,4952
MDL No.: MFCD00011060
Formula:
In₂O₃ Melting Pt: 850 °C volat. Density: 7.18 Storage Temperature: Ambient |
MDL Number:
MFCD00011060 CAS Number: 1312-43-2 EINECS: 215-193-9 Merck Index: 14,4952 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...