Indium (III) oxide ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Indium trioxide, Diindium trioxide

012544-09 012544-18 012544-30
AA12544-09EA 84.7 USD
AA12544-09 AA12544-18 AA12544-30
Indium (III) oxide ≥99.99% (metals basis)
Indium (III) oxide

MDL: MFCD00011060


Purity: 99.99% CAS No.: 1312-43-2 Molecular Formula: In2O3 Formula Weight: 277.64 Physical Form: -325 Mesh Powder Density: 7.18 Melting Point: 850° volat. Merck Reference: 14,4952 MDL No.: MFCD00011060

Formula: In₂O₃
Melting Pt: 850 °C volat.
Density: 7.18
Storage Temperature: Ambient
MDL Number: MFCD00011060
CAS Number: 1312-43-2
EINECS: 215-193-9
Merck Index: 14,4952

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR