Copper(II) nitrate hemipentahydrate 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cupric Nitrate Hydrate, Copper dinitrate hemipentahydrate, Cupric nitrate hemi(pentahydrate), Cupric nitrate 2.5-hydrate, Cupric nitrate hemipentahydrate
Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black, and producing a burnished effect on iron
Purity: 98.0-102.0%
CAS No.: 19004-19-4
Molecular Formula: CuN2O6.2.5H2O
Formula Weight: 232.59 (187.55anhy)
Physical Form: Crystalline Lumps
Density: 2.32
Appearance: Blue
Odor: Odorless
Melting Point: ca 115°
Boiling Point: 170° dec.
Solubility: Soluble in water and alcohol
Merck Reference: 14,2643
UN No.: 3085
MDL No.: MFCD00149671
Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black, and producing a burnished effect on iron
Formula:
Cu(NO₃)₂·2.5H₂O Boiling Pt: 170 °C (dec.) Melting Pt: ∼115 °C Density: 2.32 Storage Temperature: Ambient |
MDL Number:
MFCD02253001 CAS Number: 19004-19-4 EINECS: 221-838-5 UN: 3085 ADR: 5.1,III Merck Index: 14,2643 |
Specification Test Results
Insoluble matter | 0.01% |
Cl | 0.002% |
SO3 | 0.01% |
Ca | 0.005% |
Ni | 0.01% |
K | 0.005% |
NA | 0.0001 |
PB | 0.00001 |
Fe | 0.00005 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...