Copper(II) nitrate hemipentahydrate 98.0-102.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cupric Nitrate Hydrate, Copper dinitrate hemipentahydrate, Cupric nitrate hemi(pentahydrate), Cupric nitrate 2.5-hydrate, Cupric nitrate hemipentahydrate

012523-A9 012523-A3 012523-36 012523-18
AA12523-A9EA 1111 USD
AA12523-A9 AA12523-A3 AA12523-36 AA12523-18
Copper(II) nitrate hemipentahydrate 98.0-102.0% ACS
Copper (II) nitrate hemipentahydrate

Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black, and producing a burnished effect on iron


Purity: 98.0-102.0% CAS No.: 19004-19-4 Molecular Formula: CuN2O6.2.5H2O Formula Weight: 232.59 (187.55anhy) Physical Form: Crystalline Lumps Density: 2.32 Appearance: Blue Odor: Odorless Melting Point: ca 115° Boiling Point: 170° dec. Solubility: Soluble in water and alcohol Merck Reference: 14,2643 UN No.: 3085 MDL No.: MFCD00149671


Catalyst, nitrating agent for aromatic organosilicon compounds, nickel-plating baths, coloring copper black, and producing a burnished effect on iron

Formula: Cu(NO₃)₂·2.5H₂O
Boiling Pt: 170 °C (dec.)
Melting Pt: ∼115 °C
Density: 2.32
Storage Temperature: Ambient
MDL Number: MFCD02253001
CAS Number: 19004-19-4
EINECS: 221-838-5
UN: 3085
ADR: 5.1,III
Merck Index: 14,2643

Order Now

Specification Test Results

Insoluble matter 0.01%
Cl 0.002%
SO3 0.01%
Ca 0.005%
Ni 0.01%
K 0.005%
NA 0.0001
PB 0.00001
Fe 0.00005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR