Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.1 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
012421-FI
012421-FF
AA12421-FIEA
807
USD
AA12421-FI
AA12421-FF
Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness 0.1 mm (0.004 in)
Samarium
MDL: MFCD00011233
Purity: 99.9%
CAS No.: 7440-19-9
Physical Form: ≈0.47g/25x25mm, Packaged in mylar under argon
Odor: Odorless
UN No.: 3178
MDL No.: MFCD00011233
Formula:
Sm MW: 150.36 g/mol Boiling Pt: 1900 Melting Pt: 1076.85 Density: 7.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011233 CAS Number: 7440-19-9 EINECS: 231-128-7 UN: 3178 ADR: 4.1 II Merck Index: 12,08498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...