Triethylamine 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(Diethylamino)ethane, TEA
012391-30
012391-A1
012391-D5
AA12391-30EA
78.36
USD
AA12391-30
AA12391-A1
AA12391-D5
Triethylamine 99+%
Triethylamine
MDL: MFCD00009051
Beilstein Registry No.: 1843166
Fieser: 1,1198 2,427 4,527 5,689 7,385 9,481 15,336
Purity: 99+%
CAS No.: 121-44-8
Molecular Formula: C6H15N
Formula Weight: 101.19
Flash Point: -11°(12°F)
Physical Form: Liquid
Density: 0.726
Refractive Index: 1.4005
Appearance: Colorless
Odor: Ammonia-like
Melting Point: -115°
Boiling Point: 89-90°
Beilstein Reference: 1843166
Merck Reference: 14,9666
UN No.: 1296
MDL No.: MFCD00009051
Formula:
(C₂H₅)₃N Boiling Pt: 89…90 °C Melting Pt: –115 °C Density: 0.726 Flash Pt: –11 °C (–12 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009051 CAS Number: 121-44-8 EINECS: 204-469-4 UN: 1296 ADR: 3,II Merck Index: 14,9666 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...