Aluminium hydroxide ≥76.5%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hydrargillite, Aluminum hydroxide
As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmetics
Purity: 76.5% min
CAS No.: 21645-51-2
Molecular Formula: AlH3O3
Formula Weight: 78.00
Physical Form: Granular
Appearance: White
Solubility: Practically insoluble in water. Soluble in alkaline aqueous solution, HCl, H2SO4 solution
Merck Reference: 14,342
MDL No.: MFCD00003420
As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmetics
Formula:
Al(OH)₃ MW: 78 g/mol Melting Pt: 300 Density: 2.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003420 CAS Number: 21645-51-2 EINECS: 244-492-7 Merck Index: 14,342 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...