Nickel (II) oxide ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
012359-30
012359-18
AA12359-30EA
148.75
USD
AA12359-30
AA12359-18
Nickel (II) oxide ≥99% (metals basis)
Nickel (II) oxide
MDL: MFCD00011145
Purity: 99%
CAS No.: 1313-99-1
Molecular Formula: NiO
Formula Weight: 74.71
Physical Form: -325 Mesh Powder
Density: 6.67
Melting Point: 1984°
Merck Reference: 14,6512
MDL No.: MFCD00011145
Formula:
NiO Melting Pt: 1984 °C Density: 6.67 Storage Temperature: Ambient |
MDL Number:
MFCD00011145 CAS Number: 1313-99-1 EINECS: 215-215-7 Merck Index: 14,6512 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...