Magnesium chloride, anhydrous 99%
Supplier: Thermo Scientific Chemicals
Warning
|
012315-C4
012315-A7
012315-A1
AA12315-C4EA
487
USD
AA12315-C4
AA12315-A7
AA12315-A1
Magnesium chloride, anhydrous 99%
Magnesium chloride
Catalyst, flocculating agent, various magnesium cements, ceramics, analytical reagent
Purity: 99%
CAS No.: 7786-30-3
Molecular Formula: MgCl2
Formula Weight: 95.22
Physical Form: Crystalline
Density: 2.32
Appearance: White to yellow
Odor: Odorless
Melting Point: 714°
Solubility: Soluble in water with evolution of heat
Merck Reference: 14,5662
MDL No.: MFCD00011106
Catalyst, flocculating agent, various magnesium cements, ceramics, analytical reagent
Formula:
MgCl₂ MW: 95.21 g/mol Boiling Pt: 1412 °C (1013 hPa) Melting Pt: 714 °C Density: 2.32 Storage Temperature: Ambient |
MDL Number:
MFCD00011106 CAS Number: 7786-30-3 EINECS: 232-094-6 Merck Index: 14,5662 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...