Zinc chloride 97% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

012307-A7 012307-30 012307-A1
AA12307-A7EA 701 USD
AA12307-A7 AA12307-30 AA12307-A1
Zinc chloride 97% ACS
Zinc chloride

Catalyst, dehydrating and condensing agent, soldering flux, metal etchant


Purity: 97% CAS No.: 7646-85-7 Molecular Formula: Cl2Zn Formula Weight: 136.29 Physical Form: Lump Density: 2.91 Appearance: White Odor: Odorless Melting Point: 290° Boiling Point: 732° Solubility: Freely soluble in water, dilute HCl, alcohol, glycerol, acetone, ether. Zinc oxychloride forms with much water Merck Reference: 14,10132 UN No.: 2331 MDL No.: MFCD00011295


Catalyst, dehydrating and condensing agent, soldering flux, metal etchant

Formula: ZnCl₂
Boiling Pt: 732 °C
Melting Pt: 290 °C
Density: 2.91
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 14,10132

Order Now

Specification Test Results

Oxychloride Pass test
Insoluble matter 0.005%
Ca 0.06%
MG 0.01%
NO3 0.003%
SO3 0.01%
NH4 0.005%
Fe 0.001%
PB 0.005%
K 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR