Tin(IV) oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

012283-36 012283-A3 012283-22
AA12283-36EA 157 USD
AA12283-36 AA12283-A3 AA12283-22
Tin(IV) oxide ≥99.9% (metals basis)
Tin (IV) oxide

Powder, less than 10µm. Purity based on metal contaminates only.


Purity: 99.9% CAS No.: 18282-10-5 Molecular Formula: O2Sn Formula Weight: 150.69 Physical Form: 10 Micron Powder Density: 6.95 Refractive Index: 1.9968 Appearance: White to pale gray Odor: Odorless Melting Point: 1630° Boiling Point: 1800-1900° subl. Merck Reference: 14,8778 MDL No.: MFCD00011244

Formula: O₂Sn
MW: 150.71 g/mol
Boiling Pt: 1800…1900 °C (subl.)
Melting Pt: 1624.85
Density: 6.95
Storage Temperature: Ambient
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0
Merck Index: 14,08778

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR