Mercury(II) acetate ≥98.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Mercury diacetate, Mercuric acetate, Acetic acid mercury (II) salt
012273-18
012273-30
AA12273-18EA
70.7
USD
AA12273-18
AA12273-30
Mercury(II) acetate ≥98.0% ACS
Mercury (II) acetate
MDL: MFCD00012165
Beilstein Registry No.: 3563831
Fieser: 1,644 6,358 10,252 15,198 17,176 18,232 19,211 20,238 21,263
Purity: 98.0% min
CAS No.: 1600-27-7
Molecular Formula: C4H6HgO4
Formula Weight: 318.68
Physical Form: Crystalline
Density: 3.28
Appearance: White
Odor: Weak
Melting Point: 179-182°
Beilstein Reference: 3563831
Merck Reference: 14,5873
UN No.: 1629
MDL No.: MFCD00012165
Formula:
(H₃CCOO)₂Hg Melting Pt: 179…182 °C Density: 3.28 Storage Temperature: Ambient |
MDL Number:
MFCD00012165 CAS Number: 1600-27-7 EINECS: 216-491-1 UN: 1629 ADR: 6.1,II Merck Index: 14,5873 |
Specification Test Results
Insoluble matter | 0.01% |
Residue after reduction | 0.02% |
Cl | 0.005% |
NO3 | 0.005% |
SO3 | 0.005% |
Heavy Metals (as Pb) | 0.002% |
Fe | 0.001% |
Mercurous mercury (as Hg) | 0.4% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...