Strontium nitrate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00011248
Purity: 99.0% min
CAS No.: 10042-76-9
Molecular Formula: N2O6Sr
Formula Weight: 211.63
Physical Form: Crystalline
Density: 2.990
Appearance: White
Odor: Odorless
Melting Point: 570°
Merck Reference: 14,8845
UN No.: 1507
MDL No.: MFCD00011248
Formula:
Sr(NO₃)₂ Boiling Pt: 645 °C (1013 hPa) Melting Pt: 570 °C Density: 2.990 Storage Temperature: Ambient |
MDL Number:
MFCD00011248 CAS Number: 10042-76-9 EINECS: 233-131-9 UN: 1507 ADR: 5.1,III Merck Index: 14,8845 |
Specification Test Results
Insoluble matter | 0.01% |
Loss on drying at 105°C | 0.1% |
pH of a 5% solution | 5.0-7.0 at 25° |
Cl | 0.002% |
SO3 | 0.005% |
Ba | 0.05% |
Ca | 0.05% |
MG | 0.1% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 5ppm ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...