Sodium molybdate(VI) Mo 46,2%, anhydrous
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium molybdenum oxide, Sodium molybdate, Molybdic acid sodium salt, Molybdic acid, sodium salt, di-Sodium molybdate
012209-22
012209-36
AA12209-22EA
64.2
USD
AA12209-22
AA12209-36
Sodium molybdate(VI) Mo 46,2%, anhydrous
Sodium molybdate(VI)
MDL: MFCD00003486
CAS No.: 7631-95-0
Molecular Formula: MoNa2O4
Formula Weight: 205.92
Physical Form: Powder
Density: 3.72
Refractive Index: 1.714
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 687°
Merck Reference: 14,8645
MDL No.: MFCD00003486
Formula:
MoNa₂O₄ Melting Pt: 687 °C Density: 3.72 Storage Temperature: Ambient |
MDL Number:
MFCD00003486 CAS Number: 7631-95-0 EINECS: 231-551-7 Merck Index: 14,8645 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...