Strontium hydroxide octahydrate, tech.

Supplier: Thermo Scientific Chemicals
Danger

012199-18 012199-A3 012199-A9
AA12199-18EA 29.71 USD
AA12199-18 AA12199-A3 AA12199-A9
Strontium hydroxide octahydrate, tech.
Strontium hydroxide octahydrate

MDL: MFCD00150395


CAS No.: 1311-10-0 Molecular Formula: Sr(OH)2.8H2O Formula Weight: 265.76 (121.60anhy) Physical Form: Crystalline Density: 1.900 Sensitivity: Air Sensitive and Hygroscopic Appearance: White Odor: Odorless Melting Point: 100° -8H2O Merck Reference: 14,8843 UN No.: 3262 MDL No.: MFCD00150395

Formula: H₂O₂Sr·8H₂O
Melting Pt: 100 °C (-8H₂O)
Density: 1.900
Storage Temperature: Ambient
MDL Number: MFCD00150395
CAS Number: 1311-10-0
EINECS: 242-367-1
UN: 3262
ADR: 8,III
Merck Index: 14,8843

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR