Barium acetate 99.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
012198-30
012198-A1
012198-A7
AA12198-30EA
71.19
USD
AA12198-30
AA12198-A1
AA12198-A7
Barium acetate 99.0-102.0% ACS
Barium acetate
MDL: MFCD00012447
Purity: 99.0-102.0%
CAS No.: 543-80-6
Molecular Formula: Ba(OOCCH3)2
Formula Weight: 255.43
Physical Form: Crystalline
Density: 2.47
Merck Reference: 14,965
UN No.: 1564
MDL No.: MFCD00012447
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...