Barium nitrate 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
012194-18
012194-36
012194-A3
AA12194-18EA
85.82
USD
AA12194-18
AA12194-36
AA12194-A3
Barium nitrate 99+% ACS
Barium nitrate
MDL: MFCD00003442
Freely soluble in water. Slightly soluble in alcohol and acetone
Purity: 99+%
CAS No.: 10022-31-8
Molecular Formula: Ba(NO3)2
Formula Weight: 261.35
Physical Form: Crystalline
Density: 3.24
Melting Point: 592° dec.
Solubility: Freely soluble in water. Slightly soluble in alcohol and acetone
Merck Reference: 14,983
UN No.: 1446
MDL No.: MFCD00003442
Formula:
Ba(NO₃)₂ Melting Pt: 592 °C (dec.) Density: 3.24 Storage Temperature: Ambient |
MDL Number:
MFCD00003442 CAS Number: 10022-31-8 EINECS: 233-020-5 UN: 1446 ADR: 5.1,II Merck Index: 14,983 |
Specification Test Results
Insoluble matter | 0.01% |
pH of a 5 % solution | 5.0-8.0 at 25° |
Cl | 5 ppm |
K | 0.005% |
NA | 0.005% |
Ca | 0.05% |
Heavy Metals (as Pb) | 5 ppm max. |
Fe | 2 ppm |
Sr | 0.1%% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...