Potassium hexachloroplatinate(IV) Pt 39,6%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dipotassium hexachloroplatinate

012169-01 012169-03 012169-06
AA12169-01EA 47.3 USD
AA12169-01 AA12169-03 AA12169-06
Potassium hexachloroplatinate(IV) Pt 39,6%
Potassium hexachloroplatinate(IV)

MDL: MFCD00011389 Soluble in hot water. Slightly soluble in cold water. Practically insoluble in alcohol


CAS No.: 16921-30-5 Molecular Formula: Cl6K2Pt Formula Weight: 486.01 Physical Form: Crystalline powder Density: 3.50 Refractive Index: ≈1.825 Sensitivity: Hygroscopic Appearance: Yellow Odor: Odorless Melting Point: 250°dec. Solubility: Soluble in hot water. Slightly soluble in cold water. Practically insoluble in alcohol Merck Reference: 14,7636 UN No.: 3288 MDL No.: MFCD00011389

Formula: K₂PtCl₆
Melting Pt: 250 °C. (dec.)
Density: 3.50
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 3288
ADR: 6.1,III
Merck Index: 14,7636

Order Now

Specification Test Results

Form Crystalline powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR