Platinum, black
Supplier: Thermo Scientific Chemicals
012076-06
012076-77
012076-03
AA12076-06EA
935
USD
AA12076-06
AA12076-77
AA12076-03
Platinum, black
Platinum
MDL: MFCD00011179
CAS No.: 7440-06-4
Physical Form: S.A. 10m2/g
Odor: Odorless
MDL No.: MFCD00011179
Formula:
Pt Boiling Pt: 4530 °C (1013 hPa) Melting Pt: 1773 °C Density: 21.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 EINECS: 231-116-1 UN: 3089 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...