Platinum, black

Supplier: Thermo Scientific Chemicals

012076-06 012076-77 012076-03
AA12076-06EA 935 USD
AA12076-06 AA12076-77 AA12076-03
Platinum, black
Platinum

MDL: MFCD00011179


CAS No.: 7440-06-4 Physical Form: S.A. 10m2/g Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
Boiling Pt: 4530 °C (1013 hPa)
Melting Pt: 1773 °C
Density: 21.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR