Zinc ≥99.9985% (metals basis), wire, Puratronic®, Diameter 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
012053-CB
012053-BW
AA12053-CBEA
262
USD
AA12053-CB
AA12053-BW
AA12053-CP
Zinc ≥99.9985% (metals basis), wire, Puratronic®, Diameter 1.0 mm (0.04 in)
Zinc
MDL: MFCD00011291
Purity: 99.9985%
CAS No.: 7440-66-6
Physical Form: ≈5.60g/m
Odor: Odorless
MDL No.: MFCD00011291
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 Melting Pt: 420 Density: 7.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...