Zinc ≥99.9985% (metals basis), foil, Puratronic®, Thickness 1.0 mm (0.04 in)
Supplier: Thermo Scientific Chemicals
011915-FI
011915-GH
011915-FF
AA11915-FIEA
311
USD
AA11915-FI
AA11915-GH
AA11915-FF
Zinc ≥99.9985% (metals basis), foil, Puratronic®, Thickness 1.0 mm (0.04 in)
Zinc
MDL: MFCD00011291
Purity: 99.9985%
CAS No.: 7440-66-6
Physical Form: ≈4.5g/25x25mm
Odor: Odorless
MDL No.: MFCD00011291
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 Melting Pt: 420 Density: 7.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...