Zinc ≥99.994% (metals basis), foil, Puratronic®, Thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

011913-FI 011913-GH
AA11913-FIEA 127.86 USD
AA11913-FI AA11913-GH
Zinc ≥99.994% (metals basis), foil, Puratronic®, Thickness 0.25 mm (0.01 in)
Zinc

MDL: MFCD00011291


Purity: 99.994% CAS No.: 7440-66-6 Physical Form: ≈4.5g/50x50mm Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR