Tin ≥99.998% (metals basis), wire, Puratronic®, Diameter 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

011905-BW 011905-CB 011905-CD
AA11905-BWEA 38.86 USD
AA11905-BW AA11905-CB AA11905-CD
Tin ≥99.998% (metals basis), wire, Puratronic®, Diameter 1.0 mm (0.04 in)
Tin

MDL: MFCD00133862


Purity: 99.998% CAS No.: 7440-31-5 Physical Form: ≈5.72g/m Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 23-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR